Assay Details
| Gene Symbol: Chfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289577.1 | NP_001276506.1 | 12 - 13 | 1649 | 79 | |
| NM_001289578.1 | NP_001276507.1 | 11 - 12 | 1433 | 79 | ||
| NM_001289579.1 | NP_001276508.1 | 13 - 14 | 1722 | 79 | ||
| NM_001289580.1 | NP_001276509.1 | 13 - 14 | 1816 | 79 | ||
| NM_172717.4 | NP_766305.2 | 12 - 13 | 1646 | 79 | ||
| GenBank mRNA | AK048930.1 | - | 13 - 14 | 1797 | 79 | |
| AK077629.1 | - | 12 - 13 | 1618 | 79 | ||
| AK133154.1 | - | 12 - 13 | 1546 | 79 | ||
| AK147892.1 | - | 11 - 12 | 1408 | 79 | ||
| AK148436.1 | - | 12 - 13 | 1620 | 79 | ||
| AK154038.1 | - | 12 - 13 | 1631 | 79 | ||
| AK155295.1 | - | 12 - 13 | 1618 | 79 | ||
| AK155525.1 | - | 12 - 13 | 1618 | 79 | ||
| AK169182.1 | - | 13 - 14 | 1675 | 79 | ||
| AK169193.1 | - | 12 - 13 | 1601 | 79 | ||
| BC049792.1 | - | 12 - 13 | 1603 | 6510480 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chfr
Genomic Map