Assay Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162965.1 | NP_001156437.1 | 10 - 11 | 1090 | 80 | |
| NM_001254731.1 | NP_001241660.1 | 10 - 11 | 1090 | 80 | ||
| NM_172718.3 | NP_766306.2 | 10 - 11 | 1090 | 80 | ||
| NR_027934.1 | 1 - 2 | 277 | 80 | |||
| NR_027936.1 | 1 - 2 | 277 | 80 | |||
| GenBank mRNA | AK044716.1 | - | 10 - 11 | 1044 | 80 | |
| AK051018.1 | - | 1 - 2 | 278 | 80 | ||
| AK053555.1 | - | 10 - 11 | 1024 | 80 | ||
| AK170021.1 | - | 10 - 11 | 1082 | 80 | ||
| BC138049.1 | - | 10 - 11 | 1016 | 8861068 | 80 | |
| BC138050.1 | - | 10 - 11 | 1016 | 8861069 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm1
Genomic Map