Assay Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162965.1 | NP_001156437.1 | 17 - 18 | 2048 | 72 | |
| NM_001254731.1 | NP_001241660.1 | 17 - 18 | 2048 | 72 | ||
| NM_172718.3 | NP_766306.2 | 17 - 18 | 2048 | 72 | ||
| NR_027934.1 | 8 - 9 | 1235 | 72 | |||
| NR_027936.1 | 8 - 9 | 1235 | 72 | |||
| GenBank mRNA | AK044716.1 | - | 17 - 18 | 2002 | 72 | |
| AK051018.1 | - | 8 - 9 | 1236 | 72 | ||
| AK053555.1 | - | 17 - 18 | 1982 | 72 | ||
| AK167632.1 | - | 8 - 9 | 1199 | 72 | ||
| AK170021.1 | - | 17 - 18 | 2040 | 72 | ||
| BC138049.1 | - | 17 - 18 | 1974 | 8861068 | 72 | |
| BC138050.1 | - | 17 - 18 | 1974 | 8861069 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm1
Genomic Map