Assay Details
| Gene Symbol: Brat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276287.1 | NP_001263216.1 | 4 - 5 | 611 | 86 | |
| NM_181066.3 | NP_851411.1 | 4 - 5 | 513 | 86 | ||
| NR_074085.1 | 4 - 5 | 513 | 86 | |||
| GenBank mRNA | AK043212.1 | - | 4 - 5 | 476 | 86 | |
| AK049321.1 | - | 4 - 5 | 502 | 86 | ||
| AK084987.1 | - | 4 - 5 | 502 | 86 | ||
| AK085091.1 | - | 4 - 5 | 502 | 86 | ||
| AK142387.1 | - | 3 - 4 | 1686 | 86 | ||
| AK154448.1 | - | 4 - 5 | 484 | 86 | ||
| AK170269.1 | - | 4 - 5 | 506 | 86 | ||
| BC068301.1 | - | 4 - 5 | 577 | 30061848 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK170269.1 transcripts.
Set Membership: |
|

Brat1
Genomic Map