Assay Details
| Gene Symbol: Brat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276287.1 | NP_001263216.1 | 6 - 7 | 1107 | 93 | |
| NM_181066.3 | NP_851411.1 | 6 - 7 | 1009 | 93 | ||
| GenBank mRNA | AK043212.1 | - | 6 - 7 | 996 | 93 | |
| AK084987.1 | - | 6 - 7 | 998 | 93 | ||
| AK085091.1 | - | 6 - 7 | 998 | 93 | ||
| AK154448.1 | - | 6 - 7 | 980 | 93 | ||
| AK170269.1 | - | 6 - 7 | 1003 | 93 | ||
| BC068301.1 | - | 6 - 7 | 1073 | 30061848 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brat1
Genomic Map