Assay Details
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172728.2 | NP_766316.1 | 1 - 2 | 66 | 72 | |
| GenBank mRNA | AK052230.1 | - | 1 - 2 | 69 | 72 | |
| AK052451.1 | - | 1 - 2 | 67 | 72 | ||
| AK154148.1 | - | 1 - 2 | 67 | 72 | ||
| AK155537.1 | - | 1 - 2 | 64 | 72 | ||
| AK170128.1 | - | 1 - 2 | 67 | 72 | ||
| BC098487.1 | - | 1 - 2 | 56 | 30090687 | 72 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK052451.1,AK154148.1,AK155537.1,BC098487.1,NM_172728.2 transcripts.
Set Membership: |
|

Creb5
Genomic Map