Assay Details
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172728.2 | NP_766316.1 | 5 - 6 | 854 | 72 | |
| GenBank mRNA | AK052451.1 | - | 5 - 6 | 855 | 72 | |
| AK154148.1 | - | 5 - 6 | 855 | 72 | ||
| AK155537.1 | - | 5 - 6 | 852 | 72 | ||
| BC029022.1 | - | 4 - 5 | 789 | 5012117 | 72 | |
| BC098487.1 | - | 5 - 6 | 844 | 30090687 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb5
Genomic Map