Assay Details
| Gene Symbol: Lrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172814.3 | NP_766402.1 | 2 - 3 | 212 | 92 | |
| GenBank mRNA | AK050512.1 | - | 2 - 3 | 213 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrp12
Genomic Map