Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172858.2 | NP_766446.2 | 2 - 3 | 461 | 125 | |
| GenBank mRNA | AK032593.1 | - | 2 - 3 | 434 | 125 | |
| AK079080.1 | - | 3 - 4 | 554 | 125 | ||
| AK163530.1 | - | 2 - 3 | 269 | 125 | ||
| BC099417.1 | - | 2 - 3 | 454 | 125 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK032593.1,AK079080.1,AK163530.1,BC099417.1,NM_172858.2 transcripts.
Set Membership: |
|

Pak7
Genomic Map