Assay Details
| Gene Symbol: Slc7a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172861.3 | NP_766449.1 | 1 - 2 | 153 | 65 | |
| GenBank mRNA | AK044448.1 | - | 1 - 2 | 146 | 65 | |
| AK173217.1 | - | 1 - 2 | 129 | 65 | ||
| BC061928.1 | - | 1 - 2 | 129 | 5359765 | 65 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK044448.1,BC061928.1,NM_172861.3 transcripts.
Set Membership: |
|

Slc7a14
Genomic Map