Assay Details
| Gene Symbol: Slc7a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172861.3 | NP_766449.1 | 3 - 4 | 843 | 80 | |
| GenBank mRNA | AK044448.1 | - | 3 - 4 | 836 | 80 | |
| AK163358.1 | - | 3 - 4 | 887 | 80 | ||
| AK173217.1 | - | 3 - 4 | 819 | 80 | ||
| BC061928.1 | - | 3 - 4 | 819 | 5359765 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a14
Genomic Map