Assay Details
| Gene Symbol: Klhl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172871.2 | NP_766459.2 | 1 | 500 | 90 | |
| GenBank mRNA | AK033231.1 | - | 1 | 458 | 90 | |
| AK049803.1 | - | 1 | 455 | 90 | ||
| AK129338.1 | - | 1 | 402 | 90 | ||
| BC094556.1 | - | 1 | 442 | 6811241 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049803.1,BC094556.1,NM_172871.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091881 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154714 | AK129338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154714 | NM_172871.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391384 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106664 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154714 | AK129338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154714 | NM_172871.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091888 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106664 | - | - |
| Whole Mouse Genome G4122A | A_52_P567839 | AK049803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P567839 | AK129338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P567839 | NM_172871.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1083725 | AK144774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83615 | AK049803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83615 | AK129338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83615 | BC094556.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83615 | AK033231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P83615 | NM_172871.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113824 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106664 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03296779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03917225 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03917231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03296774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03917226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03917223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03296760 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03917227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03296759 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23091879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03296759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03388987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03364800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03917222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03430420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03364799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03388988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091912 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10514322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10505840 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433442_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10514319 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10505839 | - | - |
| Mouse Genome 430 2.0 Array | 1433442_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433442_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242521_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242521_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400024349.mm.1 | NM_172871.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024349.mm.1 | NM_172871.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024349.mm.1 | TC0400003016.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400024347.mm.1 | NM_172871.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024347.mm.1 | NM_172871.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024347.mm.1 | TC0400003016.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1433442_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433442_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1660068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660068 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592779 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246060 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246060 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592779 | - | - |
| mouse-6 v1.1 (Illumina) | scl00242521.1_200-S | - | - |
| mouse-6 v1.1 (Illumina) | scl24128.1.6_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660068 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00242521.1_200-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24128.1.6_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660068 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2592779 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1246060 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246060 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592779 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24128.1.6_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00242521.1_200-S | - | - |

Klhl9
Genomic Map