Assay Details
| Gene Symbol: Styk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172891.2 | NP_766479.1 | 6 - 7 | 1796 | 68 | |
| GenBank mRNA | AK033606.1 | - | 6 - 7 | 1796 | 68 | |
| AK035247.1 | - | 6 - 7 | 957 | 68 | ||
| AY563053.1 | - | 6 - 7 | 951 | 68 | ||
| BC150724.1 | - | 6 - 7 | 1004 | 9087635 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Styk1
Genomic Map