Assay Details
| Gene Symbol: Fam71e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172895.3 | NP_766483.3 | 10 - 11 | 2482 | 80 | |
| GenBank mRNA | AK076650.1 | - | 10 - 11 | 2541 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025891 | AK076650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025891 | NM_172895.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492037 | AK076650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492037 | NM_172895.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492037 | AK076650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492037 | NM_172895.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495109 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162842 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102239 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025891 | AK076650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025891 | NM_172895.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492037 | AK076650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492037 | NM_172895.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492037 | AK076650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492037 | NM_172895.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950404 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475894 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06114471 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102239 | - | - |
| Whole Mouse Genome G4122A | A_51_P492037 | AK076650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492037 | NM_172895.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492037 | AK076650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492037 | NM_172895.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122619 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102239 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06258396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06393096 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06258412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05999774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06258415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06393099 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06393090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06091261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06234039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06393102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06558710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06213147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06110016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950365 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559666 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559660 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559663 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559657 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019830.mm.1 | NM_172895.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019830.mm.1 | NM_172895.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019830.mm.1 | TC0700002170.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 489325 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3290609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3290609 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003290609 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233459 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233459 | - | - |
| mouse-6 v1.1 (Illumina) | scl31803.11.1_75-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3290609 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31803.11.1_75-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3290609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2700002 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2700002 | - | - |

Fam71e2
Genomic Map