Assay Details
| Gene Symbol: Dlgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145965.1 | NP_001139437.1 | 11 - 12 | 2550 | 60 | |
| NM_172910.3 | NP_766498.2 | 12 - 13 | 2649 | 60 | ||
| GenBank mRNA | AK032564.1 | - | 12 - 13 | 2650 | 60 | |
| AK033077.1 | - | 4 - 5 | 700 | 60 | ||
| AK082945.1 | - | 10 - 11 | 2422 | 60 | ||
| AY243847.3 | - | 6 - 7 | 2019 | 60 | ||
| BC118969.2 | - | 11 - 12 | 2336 | 40097764 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap2
Genomic Map