Assay Details
| Gene Symbol: Sept10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024910.3 | NP_001020081.2 | 6 - 7 | 1214 | 80 | |
| NM_001024911.2 | NP_001020082.1 | 6 - 7 | 1214 | 80 | ||
| GenBank mRNA | AK076569.1 | - | 6 - 7 | 1215 | 80 | |
| BC053752.1 | - | 6 - 7 | 828 | 6506619 | 80 | |
More Information
Set Membership: |
|

Sept10
Genomic Map