Assay Details
| Gene Symbol: Fbxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172988.4 | NP_766576.1 | 7 - 8 | 1706 | 90 | |
| NR_104379.1 | 7 - 8 | 1706 | 90 | |||
| NR_104380.1 | 7 - 8 | 1669 | 90 | |||
| GenBank mRNA | AK032391.1 | - | 7 - 8 | 1685 | 90 | |
| AK051429.1 | - | 7 - 8 | 1657 | 90 | ||
| AK083661.1 | - | 7 - 8 | 1643 | 90 | ||
| BC033265.1 | - | 7 - 8 | 1633 | 3983942 | 90 | |
More Information
Set Membership: |
|

Fbxl4
Genomic Map