Assay Details
| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172994.2 | NP_766582.1 | 3 - 4 | 584 | 86 | |
| GenBank mRNA | AK032023.1 | - | 3 - 4 | 487 | 86 | |
| AK049830.1 | - | 3 - 4 | 470 | 86 | ||
| AY348866.1 | - | 3 - 4 | 586 | 86 | ||
| BC059811.1 | - | 3 - 4 | 584 | 6849535 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r2c
Genomic Map