Assay Details
| Gene Symbol: Ip6k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173027.2 | NP_766615.1 | 2 - 3 | 403 | 82 | |
| GenBank mRNA | AK052857.1 | - | 2 - 3 | 404 | 82 | |
| BC141263.1 | - | 2 - 3 | 297 | 9055875 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ip6k3
Genomic Map