Assay Details
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034533.1 | - | 8 - 9 | 952 | 126 | |
| AK052841.1 | - | 8 - 9 | 964 | 126 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034533.1 | - | 8 - 9 | 952 | 126 | |
| AK052841.1 | - | 8 - 9 | 964 | 126 | ||
Set Membership: |
|