Assay Details
| Gene Symbol: Cbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173047.3 | NP_766635.1 | 2 - 3 | 503 | 66 | |
| GenBank mRNA | AK003232.1 | - | 2 - 3 | 508 | 66 | |
| AK028150.1 | - | 2 - 3 | 479 | 66 | ||
| BC028763.1 | - | 2 - 3 | 513 | 1313642 | 66 | |
| BC087735.1 | - | 2 - 3 | 464 | 5369996 | 66 | |
| BC096658.1 | - | 2 - 3 | 473 | 5145435 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977038 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45766 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156825 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30977051 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156825 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08688653 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03260205 | - | - |
| Whole Mouse Genome G4122A | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | BC028763.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | BC087735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | BC096658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | CF545530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481159 | AK003232.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132588 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156825 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03260205 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12896947 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12638928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30977036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30977045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13014712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12549285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12554953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12638494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12667077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12896951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12718664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12655079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12756780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12643419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12561994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13011069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12896947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30977039 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10436982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436981 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427912_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10436978 | - | - |
| Mouse Genome 430 2.0 Array | 1427912_at | - | - |
| Mouse Genome 430 2.0 Array | 1427912_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109857_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 109857_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600008095.mm.1 | NM_173047.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008095.mm.1 | NM_173047.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600008095.mm.1 | TC1600001045.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427912_PM_at | - | - |
| Mouse Expression Set 430 | 1427912_at | - | - |
| Murine Genome U74v2 Set | 161042_PM_at | - | - |
| Murine Genome U74v2 Set | 161042_at | - | - |
| Murine Genome U74v2 Set | 168153_i_at | - | - |
| Murine Genome U74v2 Set | 168153_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427912_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427912_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427912_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427912_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 803271 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6590148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2608811 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2608811 | - | - |
| mouse-6 v1.1 (Illumina) | scl48854.3.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48854.3.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2896768 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2608811 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2608811 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2896768 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl48854.3.1_11-S | - | - |

Cbr3
Genomic Map