Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161851.2 | NP_001155323.1 | 3 - 4 | 493 | 104 | |
| NM_173401.5 | NP_775577.2 | 3 - 4 | 770 | 104 | ||
| NR_104374.1 | 3 - 4 | 479 | 104 | |||
| NR_104375.1 | 2 - 3 | 418 | 104 | |||
| GenBank mRNA | AK019407.1 | - | 3 - 4 | 525 | 104 | |
| AK077452.1 | - | 3 - 4 | 556 | 104 | ||
| AK170958.1 | - | 3 - 4 | 453 | 104 | ||
| AK191511.1 | - | 2 - 3 | 366 | 104 | ||
| AK218805.1 | - | 2 - 3 | 299 | 104 | ||
| BC028884.1 | - | 3 - 4 | 562 | 4165174 | 104 | |
| CX202860.1 | - | 2 - 3 | 194 | 104 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fbxo44
Genomic Map