Assay Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161851.2 | NP_001155323.1 | 4 - 5 | 585 | 61 | |
| NM_173401.5 | NP_775577.2 | 4 - 5 | 862 | 61 | ||
| NR_104374.1 | 4 - 5 | 571 | 61 | |||
| NR_104375.1 | 3 - 4 | 510 | 61 | |||
| GenBank mRNA | AK019407.1 | - | 4 - 5 | 617 | 61 | |
| AK077452.1 | - | 4 - 5 | 648 | 61 | ||
| AK170958.1 | - | 4 - 5 | 545 | 61 | ||
| BC028884.1 | - | 4 - 5 | 654 | 4165174 | 61 | |
| CA322196.1 | - | 1 - 2 | 41 | 61 | ||
| CX202860.1 | - | 3 - 4 | 286 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo44
Genomic Map