Assay Details
| Gene Symbol: Dcun1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173408.3 | NP_775584.1 | 1 - 2 | 206 | 134 | |
| GenBank mRNA | AK029043.1 | - | 1 - 2 | 152 | 134 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK029043.1,NM_173408.3 transcripts.
Set Membership: |
|

Dcun1d3
Genomic Map