Assay Details
| Gene Symbol: Pskh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173432.2 | NP_775608.1 | 1 - 2 | 70 | 122 | |
| GenBank mRNA | AK053397.1 | - | 1 - 2 | 104 | 122 | |
| AK153861.1 | - | 1 - 2 | 117 | 122 | ||
| AK154570.1 | - | 1 - 2 | 100 | 122 | ||
| AK166338.1 | - | 1 - 2 | 93 | 122 | ||
| BC050128.1 | - | 1 - 2 | 70 | 30024345 | 122 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK053397.1,AK153861.1,AK154570.1,BC050128.1,NM_173432.2 transcripts.
Set Membership: |
|

Pskh1
Genomic Map