Assay Details
| Gene Symbol: Gimap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243199.1 | NP_001230128.1 | 2 - 3 | 113 | 99 | |
| NM_001243200.1 | NP_001230129.1 | 2 - 3 | 113 | 99 | ||
| NM_001243201.1 | NP_001230130.1 | 2 - 3 | 113 | 99 | ||
| NM_174990.4 | NP_778155.2 | 2 - 3 | 113 | 99 | ||
| NM_175048.3 | NP_778213.2 | 2 - 3 | 113 | 99 | ||
| GenBank mRNA | AA543119.1 | - | 2 - 3 | 87 | 99 | |
| AK088228.1 | - | 2 - 3 | 110 | 99 | ||
| BC005577.1 | - | 2 - 3 | 103 | 3968418 | 99 | |
| BC098226.1 | - | 1 - 2 | 88 | 99 | ||
| BY214550.1 | - | 2 - 3 | 114 | 99 | ||
| CA576036.1 | - | 2 - 3 | 101 | 99 | ||
| CB232465.1 | - | 2 - 3 | 144 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005577.1, NM_001243199.1, NM_001243200.1, NM_001243201.1, NM_174990.4, NM_175048.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976387 | CA576106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976387 | NM_174990.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976387 | BC005577.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147831 | NM_174990.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147831 | NM_175048.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147831 | AK088228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147831 | BC005577.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2147831 | BC098226.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976376 | NM_174990.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68110 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153301 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976376 | NM_174990.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976387 | CA576106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976387 | NM_174990.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976387 | BC005577.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147831 | NM_174990.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147831 | NM_175048.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147831 | AK088228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147831 | BC005577.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2147831 | BC098226.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24428772 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05890901 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05890891 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153301 | - | - |
| Whole Mouse Genome G4122A | A_52_P205001 | CA576106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P205001 | CB232465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P205001 | NM_174990.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P205001 | NM_175048.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P205001 | AK088228.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135939 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153301 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05890901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05459606 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05459602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05459605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05459603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24428860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05118061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05252331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05201654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05048364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05333166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05340691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05075955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05767438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05767441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05252310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05767427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05459603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05767439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05252317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05252334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05048367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05340684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05459605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05767435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05048361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05075959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05252313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05767436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05237837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24428750 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10538134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538133 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424375_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424374_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10538126 | - | - |
| Mouse Genome 430 2.0 Array | 1424375_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424375_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1424374_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424374_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107526_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107526_at | - | - |
| Mouse Expression Set 430 | 1424374_at | - | - |
| Mouse Expression Set 430 | 1424375_PM_s_at | - | - |
| Mouse Expression Set 430 | 1424374_PM_at | - | - |
| Mouse Expression Set 430 | 1424375_s_at | - | - |
| Murine Genome U74v2 Set | 96172_at | - | - |
| Murine Genome U74v2 Set | 96172_PM_at | - | - |
| Murine Genome U74v2 Set | 135687_PM_r_at | - | - |
| Murine Genome U74v2 Set | 135687_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424374_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424375_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424374_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424375_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424374_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424375_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424375_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424374_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 612867 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6550041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5570673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2757526 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225854 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2757526 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225854 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001202.1_249-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0107526.6_57-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550041 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0107526.6_57-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001202.1_249-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550041 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225854 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2757526 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225854 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757526 | - | - |

Gimap4
Genomic Map