Assay Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175006.2 | NP_778171.2 | 4 - 5 | 1148 | 88 | |
| GenBank mRNA | AK042530.1 | - | 1 - 2 | 126 | 88 | |
| AK083772.1 | - | 4 - 5 | 1148 | 88 | ||
| BC094915.1 | - | 4 - 5 | 1163 | 6435959 | 88 | |
| BC138151.1 | - | 4 - 5 | 1070 | 8861170 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pou6f2
Genomic Map