Assay Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175006.2 | NP_778171.2 | 6 - 7 | 1531 | 85 | |
| GenBank mRNA | AK083772.1 | - | 6 - 7 | 1531 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pou6f2
Genomic Map