Assay Details
| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301721.1 | NP_001288650.1 | 2 - 3 | 188 | 112 | |
| NM_001301722.1 | NP_001288651.1 | 2 - 3 | 178 | 112 | ||
| NM_175015.3 | NP_778180.1 | 2 - 3 | 178 | 112 | ||
| GenBank mRNA | AK077930.1 | - | 2 - 3 | 147 | 112 | |
| AK087955.1 | - | 2 - 3 | 155 | 112 | ||
| AW554443.2 | - | 3 - 4 | 112 | |||
| BC116218.2 | - | 1 - 2 | 108 | 40091232 | 112 | |
| CF104340.1 | - | 2 - 3 | 174 | 112 | ||
| CX208968.1 | - | 2 - 3 | 145 | 112 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK077930.1,AK087955.1,BC116218.2,NM_001301721.1,NM_001301722.1,NM_175015.3 transcripts.
Set Membership: |
|

Atp5g3
Genomic Map