Assay Details
| Gene Symbol: Atg4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145967.1 | NP_001139439.1 | 6 - 7 | 985 | 84 | |
| NM_175029.3 | NP_778194.3 | 6 - 7 | 1367 | 84 | ||
| GenBank mRNA | AK134838.1 | - | 6 - 7 | 1134 | 84 | |
| AK156463.1 | - | 6 - 7 | 1137 | 84 | ||
| AK170747.1 | - | 6 - 7 | 1124 | 84 | ||
| BC058981.1 | - | 6 - 7 | 1367 | 6827140 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg4c
Genomic Map