Assay Details
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175088.5 | NP_780297.3 | 2 - 3 | 481 | 134 | |
| GenBank mRNA | AK040675.1 | - | 1 - 2 | 193 | 134 | |
| AK048821.1 | - | 2 - 3 | 421 | 134 | ||
| AK080178.1 | - | 2 - 3 | 431 | 134 | ||
| AK086117.1 | - | 2 - 3 | 450 | 136 | ||
| AK134050.1 | - | 2 - 3 | 422 | 134 | ||
| AK165016.1 | - | 2 - 3 | 482 | 134 | ||
| BB603899.2 | - | 2 - 3 | 448 | 134 | ||
| BC067006.1 | - | 2 - 3 | 212 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdfic
Genomic Map