Assay Details
| Gene Symbol: Trmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175113.3 | NP_780322.2 | 2 - 3 | 382 | 72 | |
| GenBank mRNA | AK014359.1 | - | 2 - 3 | 379 | 72 | |
| AK017672.1 | - | 2 - 3 | 324 | 72 | ||
| AK028755.1 | - | 2 - 3 | 382 | 72 | ||
| AK129300.1 | - | 2 - 3 | 349 | 72 | ||
| AK164396.1 | - | 2 - 3 | 315 | 72 | ||
| AK167222.1 | - | 2 - 3 | 348 | 72 | ||
| BC027565.1 | - | 2 - 3 | 288 | 72 | ||
| BC052648.1 | - | 2 - 3 | 300 | 30030801 | 72 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK014359.1 transcripts.
Set Membership: |
|

Trmt6
Genomic Map