Assay Details
| Gene Symbol: Trmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175113.3 | NP_780322.2 | 8 - 9 | 1230 | 95 | |
| GenBank mRNA | AK014359.1 | - | 8 - 9 | 1346 | 95 | |
| AK028755.1 | - | 8 - 9 | 1230 | 95 | ||
| AK129300.1 | - | 8 - 9 | 1197 | 95 | ||
| AK165976.1 | - | 8 - 9 | 1182 | 95 | ||
| AK167222.1 | - | 8 - 9 | 1196 | 95 | ||
| BC049083.1 | - | 4 - 5 | 1358 | 95 | ||
| BC052648.1 | - | 8 - 9 | 1148 | 30030801 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt6
Genomic Map