Assay Details
| Gene Symbol: Sptlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175467.3 | NP_780676.1 | 7 - 8 | 1038 | 77 | |
| GenBank mRNA | AK048374.1 | - | 6 - 7 | 899 | 77 | |
| AK054240.1 | - | 6 - 7 | 1125 | 77 | ||
| AK078679.1 | - | 6 - 7 | 1102 | 77 | ||
| AK078686.1 | - | 7 - 8 | 1039 | 77 | ||
| BC094496.1 | - | 6 - 7 | 884 | 3483569 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc3
Genomic Map