Assay Details
| Gene Symbol: Sptlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175467.3 | NP_780676.1 | 10 - 11 | 1486 | 110 | |
| GenBank mRNA | AK048374.1 | - | 9 - 10 | 1347 | 110 | |
| AK054240.1 | - | 9 - 10 | 1573 | 110 | ||
| AK078679.1 | - | 9 - 10 | 1550 | 110 | ||
| AK078686.1 | - | 10 - 11 | 1487 | 110 | ||
| BC094496.1 | - | 9 - 10 | 1331 | 3483569 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc3
Genomic Map