Assay Details
| Gene Symbol: Gpr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175490.4 | NP_780699.2 | 2 | 1685 | 61 | |
| GenBank mRNA | AK044553.1 | - | 1 | 1152 | 61 | |
| AK141614.1 | - | 2 | 1686 | 61 | ||
| AY253852.1 | - | 1 | 1392 | 61 | ||
| BC125614.1 | - | 1 | 1481 | 40130129 | 61 | |
| BC125616.1 | - | 1 | 1481 | 40130131 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681063 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107457 | BC125614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107457 | BC125616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107457 | AK141614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107457 | AY253852.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107457 | AK044553.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399539 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116720 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111892 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107457 | BC125614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107457 | BC125616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107457 | AK141614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107457 | AY253852.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107457 | AK044553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227419 | AK141614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227419 | AK044553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227419 | NM_175490.4 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111892 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116720 | - | - |
| Whole Mouse Genome G4122A | A_51_P227419 | AK141614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227419 | AK044553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227419 | NM_175490.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110178 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116720 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111892 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448880 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09568198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448885 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09568213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09448886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27681062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27681040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09305927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09568197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09386641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09568215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09568206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09199817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09199815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09196024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09568219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27681084 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10374999 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434536_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10374998 | - | - |
| Mouse Genome 430 2.0 Array | 1434536_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434536_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100002686.mm.1 | NM_175490.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002686.mm.1 | NM_175490.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002686.mm.1 | TC1100000315.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100002688.mm.1 | NM_175490.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002688.mm.1 | NM_175490.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002688.mm.1 | TC1100000315.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434536_PM_at | - | - |
| Mouse Expression Set 430 | 1434536_at | - | - |
| Murine Genome U74v2 Set | 138063_PM_at | - | - |
| Murine Genome U74v2 Set | 138063_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434536_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434536_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 362203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980056 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980056 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980056 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2767510 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161775 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161775 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2767510 | - | - |
| mouse-6 v1.1 (Illumina) | GI_47059090-S | - | - |
| mouse-6 v1.1 (Illumina) | scl41737.1.1_327-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41737.1.1_327-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161775 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242693 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161775 | - | - |

Gpr75
Genomic Map