Assay Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175543.3 | NP_780752.1 | 6 - 7 | 956 | 61 | |
| GenBank mRNA | AK046734.1 | - | 6 - 7 | 958 | 61 | |
| AK147219.1 | - | 6 - 7 | 1027 | 61 | ||
| AK147344.1 | - | 6 - 7 | 1152 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip4
Genomic Map