Assay Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175543.3 | NP_780752.1 | 11 - 12 | 1416 | 62 | |
| GenBank mRNA | AK046734.1 | - | 11 - 12 | 1418 | 62 | |
| AK122559.1 | - | 3 - 4 | 252 | 62 | ||
| AK147219.1 | - | 11 - 12 | 1487 | 62 | ||
| AK147344.1 | - | 11 - 12 | 1612 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip4
Genomic Map