Assay Details
| Gene Symbol: Acta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272041.1 | NP_001258970.1 | 6 - 7 | 993 | 71 | |
| NM_009606.3 | NP_033736.1 | 5 - 6 | 903 | 71 | ||
| GenBank mRNA | BC014877.1 | - | 5 - 6 | 864 | 3587657 | 71 |
| M12233.1 | - | 3 - 4 | 684 | 71 | ||
| M12866.1 | - | 5 - 6 | 857 | 71 | ||
| X03766.1 | - | 3 - 4 | 684 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242187 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242184 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242182 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760826 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127554 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26242234 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127554 | - | - |
| Whole Mouse Genome G4122A | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P246854 | BC014877.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132868 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07030304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06885187 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06885184 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06885188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242182 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242181 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242187 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26242184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26242159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06881235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06927312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06842416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06839024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07030300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07160808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07146130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07030311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07194920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06881234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06839020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06881233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06881220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06842420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06839026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26242176 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10582593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10582595 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427735_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10582592 | - | - |
| Mouse Genome 430 2.0 Array | 1427735_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427735_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11459_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11459_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800024249.mm.1 | NM_001272041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024249.mm.1 | NM_009606.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024249.mm.1 | TC0800003161.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800024250.mm.1 | NM_001272041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024250.mm.1 | NM_009606.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800024250.mm.1 | TC0800003161.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427735_a_at | - | - |
| Mouse Expression Set 430 | 1427735_PM_a_at | - | - |
| Murine Genome U74v2 Set | 100381_PM_at | - | - |
| Murine Genome U74v2 Set | 100381_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427735_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427735_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427735_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427735_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916274 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 840538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 840538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000840538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2738825 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2738825 | - | - |
| mouse-6 v1.1 (Illumina) | scl34188.7.1_72-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 840538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34188.7.1_72-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 840538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2738825 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2738825 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34188.7.1_72-S | - | - |

Acta1
Genomic Map