Assay Details
| Gene Symbol: Cdc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243769.1 | NP_001230698.1 | 5 - 6 | 649 | 95 | |
| GenBank mRNA | CD353254.1 | - | 2 - 3 | 233 | 95 | |
| L78075.1 | - | 4 - 5 | 489 | 95 | ||
More Information
Set Membership: |
|

Cdc42
Genomic Map