Assay Details
| Gene Symbol: Dnmt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003960.4 | NP_001003960.2 | 3 - 4 | 587 | 68 | |
| NM_001003961.4 | NP_001003961.2 | 3 - 4 | 587 | 68 | ||
| NM_001003963.4 | NP_001003963.2 | 3 - 4 | 587 | 68 | ||
| NM_001122997.2 | NP_001116469.1 | 2 - 3 | 472 | 68 | ||
| NM_001271744.1 | NP_001258673.1 | 2 - 3 | 472 | 68 | ||
| NM_001271745.1 | NP_001258674.1 | 2 - 3 | 472 | 68 | ||
| NM_001271746.1 | NP_001258675.1 | 2 - 3 | 472 | 68 | ||
| NM_001271747.1 | NP_001258676.1 | 2 - 3 | 472 | 68 | ||
| NM_010068.5 | NP_034198.3 | 3 - 4 | 587 | 68 | ||
| GenBank mRNA | AF068626.2 | - | 3 - 4 | 442 | 68 | |
| AF068627.2 | - | 3 - 4 | 442 | 68 | ||
| AF068628.2 | - | 3 - 4 | 442 | 68 | ||
| AF151969.1 | - | 3 - 4 | 585 | 68 | ||
| AF151970.1 | - | 3 - 4 | 585 | 68 | ||
| AF151971.1 | - | 3 - 4 | 585 | 68 | ||
| AF151972.1 | - | 3 - 4 | 585 | 68 | ||
| AF151973.1 | - | 2 - 3 | 470 | 68 | ||
| AF151974.1 | - | 2 - 3 | 470 | 68 | ||
| AF151975.1 | - | 2 - 3 | 470 | 68 | ||
| AF151976.1 | - | 2 - 3 | 470 | 68 | ||
| AY078427.1 | - | 3 - 4 | 442 | 68 | ||
| BC105677.1 | - | 2 - 3 | 304 | 40048574 | 68 | |
| BC105922.1 | - | 2 - 3 | 304 | 40048584 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnmt3b
Genomic Map