Assay Details
| Gene Symbol: Esr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010157.3 | NP_034287.3 | 6 - 7 | 1488 | 71 | |
| GenBank mRNA | BC141075.1 | - | 5 - 6 | 1190 | 9055687 | 71 |
| BC145329.1 | - | 5 - 6 | 1190 | 9053885 | 71 | |
| U81451.1 | - | 5 - 6 | 960 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Effects of gestational diethylstilbestrol treatment on male and female gonads during early embryonic development.
- Authors
- Ikeda Y, Tanaka H, Esaki M
- Journal
- Endocrinology - Page No 3970 (Fri Aug 01 00:00:00 UTC 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -

Esr2
Genomic Map