Assay Details
| Gene Symbol: Pon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011134.3 | NP_035264.2 | 6 - 7 | 889 | 72 | |
| GenBank mRNA | BC012706.1 | - | 6 - 7 | 762 | 4158951 | 72 |
| U32684.1 | - | 6 - 7 | 771 | 72 | ||
| U72636.1 | - | 6 - 7 | 771 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pon1
Genomic Map