Assay Details
| Gene Symbol: Tenm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011855.4 | NP_035985.2 | 13 - 14 | 2349 | 94 | |
| GenBank mRNA | AB025410.1 | - | 13 - 14 | 2351 | 94 | |
| BC138860.1 | - | 12 - 13 | 2332 | 8861882 | 94 | |
| BC138861.1 | - | 12 - 13 | 2332 | 8861883 | 94 | |
More Information
Set Membership: |
|

Tenm1
Genomic Map