Assay Details
| Gene Symbol: Tenm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011855.4 | NP_035985.2 | 22 - 23 | 3805 | 146 | |
| GenBank mRNA | AB025410.1 | - | 22 - 23 | 3807 | 146 | |
| BC138860.1 | - | 21 - 22 | 3788 | 8861882 | 146 | |
| BC138861.1 | - | 21 - 22 | 3788 | 8861883 | 146 | |
More Information
Set Membership: |
|

Tenm1
Genomic Map