Assay Details
| Gene Symbol: Tenm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011855.4 | NP_035985.2 | 8 - 9 | 1457 | 102 | |
| GenBank mRNA | AB025410.1 | - | 8 - 9 | 1459 | 102 | |
| AK047423.1 | - | 10 - 11 | 2222 | 102 | ||
| BC138860.1 | - | 7 - 8 | 1440 | 8861882 | 102 | |
| BC138861.1 | - | 7 - 8 | 1440 | 8861883 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tenm1
Genomic Map