Assay Details
| Gene Symbol: Cabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013879.2 | NP_038907.1 | 3 - 4 | 328 | 88 | |
| GenBank mRNA | AB079604.1 | - | 3 - 4 | 267 | 88 | |
| AF169152.1 | - | 3 - 4 | 328 | 88 | ||
| AF169153.1 | - | 2 - 3 | 148 | 88 | ||
| AK141231.1 | - | 3 - 4 | 242 | 88 | ||
| KJ364651.1 | - | 2 - 3 | 766 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabp1
Genomic Map