Assay Details
| Gene Symbol: Ddx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017397.3 | NP_059093.3 | 2 - 3 | 534 | 74 | |
| GenBank mRNA | AF217000.1 | - | 2 - 3 | 409 | 74 | |
| AF217001.1 | - | 2 - 3 | 409 | 74 | ||
| AF220454.1 | - | 2 - 3 | 537 | 74 | ||
| AJ250027.1 | - | 2 - 3 | 409 | 74 | ||
| AK050950.1 | - | 2 - 3 | 408 | 74 | ||
| AK154518.1 | - | 2 - 3 | 413 | 74 | ||
| BC125471.1 | - | 2 - 3 | 534 | 40129986 | 74 | |
| BC137721.1 | - | 2 - 3 | 534 | 8860740 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx20
Genomic Map