Assay Details
| Gene Symbol: Hagh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159626.1 | NP_001153098.1 | 4 - 5 | 450 | 77 | |
| NM_024284.2 | NP_077246.2 | 4 - 5 | 540 | 77 | ||
| GenBank mRNA | AK147022.1 | - | 4 - 5 | 501 | 77 | |
| AK150557.1 | - | 4 - 5 | 458 | 77 | ||
| AK151231.1 | - | 4 - 5 | 458 | 77 | ||
| AK152742.1 | - | 4 - 5 | 458 | 77 | ||
| BC004749.1 | - | 4 - 5 | 483 | 3583919 | 77 | |
| BC019817.1 | - | 4 - 5 | 433 | 5123648 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hagh
Genomic Map