Assay Details
| Gene Symbol: Hagh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159626.1 | NP_001153098.1 | 6 - 7 | 656 | 72 | |
| NM_024284.2 | NP_077246.2 | 6 - 7 | 746 | 72 | ||
| GenBank mRNA | AK147022.1 | - | 6 - 7 | 707 | 72 | |
| AK150557.1 | - | 6 - 7 | 664 | 72 | ||
| AK151231.1 | - | 6 - 7 | 664 | 72 | ||
| AK152742.1 | - | 6 - 7 | 664 | 72 | ||
| AK165777.1 | - | 6 - 7 | 650 | 72 | ||
| BC004749.1 | - | 6 - 7 | 689 | 3583919 | 72 | |
| BC019817.1 | - | 6 - 7 | 639 | 5123648 | 72 | |
| BQ128470.1 | - | 3 - 4 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Hagh
Genomic Map